Anti-PLXNA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 707206]

Supplier: R & D Systems
FAB58561P
102883-110EA 513.38 USD
102883-110
Anti-PLXNA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 707206]
Antibodies
Type: Primary
Antigen: PLXNA4
Clonality: Monoclonal
Clone: 707206
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR