Anti-ICAM1 Mouse Monoclonal Antibody (Fluorescein) [clone: 141032]

Supplier: R & D Systems
FAB5831F
220017-894EA 316.71 USD
220017-894
Anti-ICAM1 Mouse Monoclonal Antibody (Fluorescein) [clone: 141032]
Antibodies
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 141032
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR