Anti-CX3CR1 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB5825G-100 FAB5825G-025
102883-104EA 561.11 USD
102883-104 102883-102
Anti-CX3CR1 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR