Anti-CX3CR1 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB5825A-100 FAB5825A-025
102883-100EA 594.37 USD
102883-100 102883-098
Anti-CX3CR1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR