Anti-SIGLEC12 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 750620]

Supplier: R & D Systems
FAB5806A
102883-096EA 594.37 USD
102883-096
Anti-SIGLEC12 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 750620]
Antibodies
Type: Primary
Antigen: SIGLEC12
Clonality: Monoclonal
Clone: 750620
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR