Anti-NT5E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 606112]

Supplier: R & D Systems
FAB5795P
220017-893EA 517.72 USD
220017-893
Anti-NT5E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 606112]
Antibodies
Type: Primary
Antigen: NT5E
Clonality: Monoclonal
Clone: 606112
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR