Anti-NT5E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 606112]

Supplier: R & D Systems
FAB5795A
220017-892EA 577.02 USD
220017-892
Anti-NT5E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 606112]
Antibodies
Type: Primary
Antigen: NT5E
Clonality: Monoclonal
Clone: 606112
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR