Anti-SELL Rat Monoclonal Antibody (Fluorescein) [clone: 95218]

Supplier: R & D Systems
FAB5761F
220017-889EA 241.51 USD
220017-889
Anti-SELL Rat Monoclonal Antibody (Fluorescein) [clone: 95218]
Antibodies
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: 95218
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR