Anti-CR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 594708]

Supplier: R & D Systems
FAB5748A
220017-885EA 513.38 USD
220017-885
Anti-CR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 594708]
Antibodies
Type: Primary
Antigen: CR1
Clonality: Monoclonal
Clone: 594708
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR