Anti-CD96 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 630612]

Supplier: R & D Systems
FAB5690P
102883-094EA 438.18 USD
102883-094
Anti-CD96 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 630612]
Antibodies
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 630612
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR