Anti-L1CAM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 555]

Supplier: R & D Systems
FAB5674P
220017-881EA 513.38 USD
220017-881
Anti-L1CAM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 555]
Antibodies
Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: 555
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR