Anti-NRP1 Goat Polyclonal Antibody (Alexa Fluor® 700)

Supplier: R & D Systems
FAB566N
102883-038EA 519.17 USD
102883-038
Anti-NRP1 Goat Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR