Anti-VCAM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: IE10]

Supplier: R & D Systems
FAB5649G
102883-028EA 501.82 USD
102883-028
Anti-VCAM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: IE10]
Antibodies
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: IE10
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR