Anti-VCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: IE10]

Supplier: R & D Systems
FAB5649A
220017-876EA 577.02 USD
220017-876
Anti-VCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: IE10]
Antibodies
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: IE10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR