Anti-FOLR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 548908]

Supplier: R & D Systems
FAB5646A
220017-874EA 614.62 USD
220017-874
Anti-FOLR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 548908]
Antibodies
Type: Primary
Antigen Symbol: FOLR1
Antigen Name: folate receptor 1 (adult)
Antigen Synonyms: FBP, FOLR
Clonality: Monoclonal
Clone: 548908
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR