Anti-SIGLEC1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 645608]

Supplier: R & D Systems
FAB5610P
102882-294EA 454.09 USD
102882-294
Anti-SIGLEC1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 645608]
Antibodies
Type: Primary
Antigen: SIGLEC1
Clonality: Monoclonal
Clone: 645608
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR