Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: GK1.5]

Supplier: R & D Systems
FAB554P-100 FAB554P-025
102882-290EA 177.88 USD
102882-290 102883-024
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: GK1.5]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR