Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: GK1.5]

Supplier: R & D Systems
FAB554N-100 FAB554N-025
102883-022EA 274.77 USD
102883-022 102883-020
Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: GK1.5]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR