Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: GK1.5]

Supplier: R & D Systems
FAB554G-025 FAB554G-100
102883-018EA 284.89 USD
102883-018 102883-016
Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: GK1.5]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR