Anti-CSF2RB2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 130705]

Supplier: R & D Systems
FAB5492A
220017-863EA 535.08 USD
220017-863
Anti-CSF2RB2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 130705]
Antibodies
Type: Primary
Antigen: CSF2RB2
Clonality: Monoclonal
Clone: 130705
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR