Anti-PLXNA2 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB5486A
102882-972EA 608.83 USD
102882-972
Anti-PLXNA2 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: PLXNA2
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR