Anti-CRLF2 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB5461F
220017-859EA 446.86 USD
220017-859
Anti-CRLF2 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: CRLF2
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR