Anti-CRLF2 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB5461A
102882-970EA 577.02 USD
102882-970
Anti-CRLF2 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CRLF2
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR