Anti-IL10RB Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 547324]

Supplier: R & D Systems
FAB53681G
102882-954EA 527.85 USD
102882-954
Anti-IL10RB Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 547324]
Antibodies
Type: Primary
Antigen: IL10RB
Clonality: Monoclonal
Clone: 547324
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR