Anti-TGFBR2 Goat Polyclonal Antibody (Alexa Fluor® 700)

Supplier: R & D Systems
FAB532N-025 FAB532N-100
102882-906EA 513.38 USD
102882-906 102882-904
Anti-TGFBR2 Goat Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR