Anti-TGFBR2 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB532A-025 FAB532A-100
102882-892EA 310.92 USD
102882-892 102882-894
Anti-TGFBR2 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR