Anti-PLXNB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 537223]

Supplier: R & D Systems
FAB53291P
102882-890EA 543.75 USD
102882-890
Anti-PLXNB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 537223]
Antibodies
Type: Primary
Antigen: PLXNB2
Clonality: Monoclonal
Clone: 537223
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR