Anti-IL4RA Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB530P-100 FAB530P-025
102882-884EA 258.86 USD
102882-884 102882-886
Anti-IL4RA Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: IL4RA
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR