Anti-IFNLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 601106]

Supplier: R & D Systems
FAB5260P
220017-848EA 527.85 USD
220017-848
Anti-IFNLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 601106]
Antibodies
Type: Primary
Antigen: IFNLR1
Clonality: Monoclonal
Clone: 601106
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR