Anti-CD24 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ML5]

Supplier: R & D Systems
FAB5247A
102882-872EA 501.82 USD
102882-872
Anti-CD24 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ML5]
Antibodies
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: ML5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR