Anti-TNFRSF18 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 108619]

Supplier: R & D Systems
FAB5241A
102882-870EA 552.43 USD
102882-870
Anti-TNFRSF18 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 108619]
Antibodies
Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: 108619
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR