Anti-IL1RL1 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB5231P-025 FAB5231P-100
102882-862EA 277.66 USD
102882-862 102882-864
Anti-IL1RL1 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: IL1RL1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR