Anti-IL1RL1 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB5231A-100 FAB5231A-025
102882-860EA 582.8 USD
102882-860 102882-858
Anti-IL1RL1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: IL1RL1
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR