Anti-CD9 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 479608]

Supplier: R & D Systems
FAB5218A
220017-842EA 389.02 USD
220017-842
Anti-CD9 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 479608]
Antibodies
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: 479608
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR