Anti-CX3CR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 528728]

Supplier: R & D Systems
FAB5204P-025 FAB5204P-100
102882-286EA 205.35 USD
102882-286 102882-288
Anti-CX3CR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 528728]
Antibodies
Type: Primary
Antigen: CX3CR1
Clonality: Monoclonal
Clone: 528728
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR