Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 745520]

Supplier: R & D Systems
FAB51561P
102882-848EA 227.05 USD
102882-848
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 745520]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 745520
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR