Anti-CD44 (pan specific) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2C5]

Supplier: R & D Systems
FAB4948P
220017-827EA 253.08 USD
220017-827
Anti-CD44 (pan specific) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2C5]
Antibodies
Type: Primary
Antigen: CD44 (pan specific)
Clonality: Monoclonal
Clone: 2C5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR