Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4G7-2E3]

Supplier: R & D Systems
FAB4867P
220017-814EA 241.51 USD
220017-814
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4G7-2E3]
Antibodies
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7-2E3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR