Anti-CD19 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 4G7-2E3]

Supplier: R & D Systems
FAB4867N
102882-816EA 339.85 USD
102882-816
Anti-CD19 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 4G7-2E3]
Antibodies
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7-2E3
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR