Anti-CD19 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 4G7-2E3]

Supplier: R & D Systems
FAB4867C
220017-812EA 389.02 USD
220017-812
Anti-CD19 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 4G7-2E3]
Antibodies
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7-2E3
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR