Anti-CD247 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A2]

Supplier: R & D Systems
FAB4841P-100 FAB4841P-025
102882-812EA 112.8 USD
102882-812 102882-814
Anti-CD247 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A2]
Antibodies
Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 17A2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR