Anti-CD247 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 17A2]

Supplier: R & D Systems
FAB4841G-025 FAB4841G-100
102882-806EA 342.74 USD
102882-806 102882-804
Anti-CD247 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 17A2]
Antibodies
Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 17A2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR