Anti-CD247 Rat Monoclonal Antibody (Fluorescein) [clone: 17A2]

Supplier: R & D Systems
FAB4841F-025 FAB4841F-100
102882-800EA 96.89 USD
102882-800 102882-802
Anti-CD247 Rat Monoclonal Antibody (Fluorescein) [clone: 17A2]
Antibodies
Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 17A2
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR