Anti-CD247 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 17A2]

Supplier: R & D Systems
FAB4841A-100 FAB4841A-025
102882-792EA 193.78 USD
102882-792 102882-794
Anti-CD247 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 17A2]
Antibodies
Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 17A2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR