Anti-ITGB8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 416922]

Supplier: R & D Systems
FAB4775A
220017-802EA 608.83 USD
220017-802
Anti-ITGB8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 416922]
Antibodies
Type: Primary
Antigen: ITGB8
Clonality: Monoclonal
Clone: 416922
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR