Anti-IL7R Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: A7R34]

Supplier: R & D Systems
FAB47742A
102882-778EA 501.82 USD
102882-778
Anti-IL7R Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: A7R34]
Antibodies
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: A7R34
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR