Anti-HRH1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 480054]

Supplier: R & D Systems
FAB4726P
220017-800EA 527.85 USD
220017-800
Anti-HRH1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 480054]
Antibodies
Type: Primary
Antigen: HRH1
Clonality: Monoclonal
Clone: 480054
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR