Anti-FLT1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 141522]

Supplier: R & D Systems
FAB4711N
102882-768EA 513.38 USD
102882-768
Anti-FLT1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 141522]
Antibodies
Type: Primary
Antigen Symbol: FLT1
Antigen Name: FMS-like tyrosine kinase 1
Antigen Synonyms: AI323757, Flt-1, VEGFR-1, VEGFR1, sFlt1
Clonality: Monoclonal
Clone: 141522
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR