Anti-IL6ST Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 125623]

Supplier: R & D Systems
FAB4681P-025 FAB4681P-100
102882-760EA 513.38 USD
102882-760 102882-758
Anti-IL6ST Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 125623]
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Monoclonal
Clone: 125623
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR