Anti-CD47 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 472603]

Supplier: R & D Systems
FAB4670P
220017-794EA 517.72 USD
220017-794
Anti-CD47 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 472603]
Antibodies
Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: 472603
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR